Orders LinkContactUsLinkComponent
TechniCloth® Flat Mop Covers
TechniCloth® Flat Mop Covers
Technicloth® floor mop wipers, 250×600 mm
Catalog # TEXWTX630
New Product
Supplier:   Texwipe
TechniCloth® Flat Mop Covers
Catalog # TEXWTX630
Supplier:  Texwipe
New Product
TechniCloth® Flat Mop Covers
TechniCloth® Flat Mop Covers
Technicloth® floor mop wipers, 250×600 mm
Price
1 037,86 €
Log in for availability
1 * 12 ST
Product Details & Documents
Mop covers (ISO Class 3 - 8) for cleaning and decontaminating floors.
  • Made without chemical binders or additives
  • Non-woven, economical polyester/cellulose for cleaning and removing residues
  • Chemically resistant, withstand exposure to common cleaning and disinfecting solutions
Specifications
  • Description:
    Technicloth® floor mop wipers
  • L×W:
    250×600 mm
  • Seq:
    3