Order Entry
Austria
Orders LinkContactUsLinkComponent
Notepad, self-adhesive, TexWrite® TexNotes

Notepad, self-adhesive, TexWrite® TexNotes

TexNotes notepad, TX5820, 76×102 mm, Light blue
Catalog # TEXWTX5820
New Product
Supplier:   Texwipe
Notepad, self-adhesive, TexWrite® TexNotes
Catalog # TEXWTX5820
Supplier:  Texwipe
New Product
Notepad, self-adhesive, TexWrite® TexNotes
Notepad, self-adhesive, TexWrite® TexNotes

TexNotes notepad, TX5820, 76×102 mm, Light blue

Price
147,38 €
Log in for availability
1 * 1 ST
Product Details & Documents

Ideal for leaving notes on documents, reports, instruments, and most cleanroom surfaces.

  • Low contamination polymeric elastomer adhesive allows TexNotes® to be attached and removed without high residue

Format: 102×76 mm

Packaging: 72 sheets per pad, 10 pads per pack (Bag-Within-A-Bag®).

Documents

Specifications
  • Colour:
    Light blue
  • Description:
    TexNotes notepad, TX5820
  • Dimensions:
    76×102 mm
  • Seq:
    2
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.