Orders LinkContactUsLinkComponent
Inserts
Inserts
A 315/1 module 492×503×118 mm (W×D×H) for increased capacity and reprocessing of lumened instruments, increases capacity from 22 to 44, can be used in all A 151 lower baskets available on the market, compatible with all standard dental and ENT accessories; Injector nozzles not included, A 800 tubular filter for use in either; Clearances in lower basket A 151: H 210 ±30, W 383, D 479 mm injector manifold (not included); For use with: PG 8581/62/82/91/92, PWA 8672, PWD 8682/92, 492×503×118 mm
Catalog # MIEL12531630
New Product
Supplier:   MIELE
Inserts
Catalog # MIEL12531630
Supplier:  MIELE
New Product
Inserts
Inserts
A 315/1 module 492×503×118 mm (W×D×H) for increased capacity and reprocessing of lumened instruments, increases capacity from 22 to 44, can be used in all A 151 lower baskets available on the market, compatible with all standard dental and ENT accessories; Injector nozzles not included, A 800 tubular filter for use in either; Clearances in lower basket A 151: H 210 ±30, W 383, D 479 mm injector manifold (not included); For use with: PG 8581/62/82/91/92, PWA 8672, PWD 8682/92, 492×503×118 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    A 315/1 module 492×503×118 mm (W×D×H) for increased capacity and reprocessing of lumened instruments, increases capacity from 22 to 44, can be used in all A 151 lower baskets available on the market, compatible with all standard dental and ENT accessories; Injector nozzles not included, A 800 tubular filter for use in either; Clearances in lower basket A 151: H 210 ±30, W 383, D 479 mm injector manifold (not included); For use with: PG 8581/62/82/91/92, PWA 8672, PWD 8682/92
  • Seq:
    57
  • W×D×H:
    492×503×118 mm