Orders LinkContactUsLinkComponent
Jars, without Cap
PP, 250 ml, —, —, 83,4 mm
  RIXI4-1101-0250-65
New Product
Jars, without Cap
  RIXI4-1101-0250-65
New Product
Jars, without Cap
Jars, without Cap
PP, 250 ml, —, —, 83,4 mm
1 * 200 ST
These slip lid jars are made of white PP.
  • Very good chemical resistance
  • Very good temperature resistance
 : Caps need to be ordered seperately.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
  • Ext.Ø:
  • Type:
    PP
  • Capacity:
    250 ml
  • Height:
    83,4 mm
  • Seq:
    4
  • Neck I-Ø:
Recommendations