Orders LinkContactUsLinkComponent
Tool carts
Tool cart, with 6 drawers, secure lock and cleanroom castors with brakes, 730×470×1000 mm
Catalog # PALBTOOLCART6
New Product
Supplier:   Palbam
Tool carts
Catalog # PALBTOOLCART6
Supplier:  Palbam
New Product
Tool carts
Tool carts
Tool cart, with 6 drawers, secure lock and cleanroom castors with brakes, 730×470×1000 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Tool cart in stainless steel 304 with electropolished finish.
  • Five fully extendable drawers
  • One lock which secures all the drawers
  • Top ledge to prevent tools/parts falling
  • Cleanroom castors with brakes
Specifications
  • Description:
    Tool cart, with 6 drawers, secure lock and cleanroom castors with brakes
  • Seq:
    2
  • W×D×H:
    730×470×1000 mm