Caffeic acid
Bottle
Catalog # APOSBIB6021-500G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
- CAS Number:331-39-5
- Formula:C₉H₈O₄
- MDL Number:MFCD00004392
- Molecular Weight:180.16 g/mol
- Melting Pt:211…213 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-361-2
Specifications
Specifications
- Pack type:Bottle



