Order Entry
Austria
Orders LinkContactUsLinkComponent
Decoquinate 98%

Decoquinate 98%

Bottle
Catalog # APOSOR1030278-2.5K
New Product
Supplier:   Apollo Scientific
Decoquinate 98%
Catalog # APOSOR1030278-2.5K
Supplier:  Apollo Scientific
New Product
CAS Number:  18507-89-6
Decoquinate 98%

Bottle

CAS Number:  18507-89-6
Price
2 950,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester
  • CAS Number:18507-89-6
  • Formula:C₂₄H₃₅NO₅
  • Molecular Weight:417,54 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications