Orders LinkContactUsLinkComponent
Dispensing Lines
Additional sets of dispensing lines, TUB-DOSIF-4, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 4 mm int.Ø, 7,2 mm ext.Ø
  ESPITUB-DOSIF-4
New Product
 :   RAYPA
Dispensing Lines
  ESPITUB-DOSIF-4
 :  RAYPA
New Product
Dispensing Lines
Dispensing Lines
Additional sets of dispensing lines, TUB-DOSIF-4, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 4 mm int.Ø, 7,2 mm ext.Ø
1 * 1 ST
Additional silicone dosing tubes, with metal nozzle for dispensing sterile culture media.
  • Silicone dosing tubes with press-fit connection and metal nozzle
  • Sterile medium is dispensed through the flame-sterilizable nozzle
  • Made of MVQ silicone; heat-resistant up to 200 °C; 60 A hardness
  • Available in multiple diameters
  • Steam and pressurised air enable easy line cleaning before, during, and after use
  • All models are autoclavable, but autoclaving after each use is unnecessary thanks to the built-in steam cleaning program
 : Dispensing lines can be customised, adjusting their length or adding modifications to adapt them to the specific production requirements of the customer.
  • Description:
    Additional sets of dispensing lines, TUB-DOSIF-4, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 4 mm int.Ø, 7,2 mm ext.Ø
  • Seq:
    2