Orders LinkContactUsLinkComponent
Temperature Data Loggers
Temperature Data Loggers
AISI-316L stainless steel disk temperature recorder DLDISK3618_CL, 80 g, for accurate and precise temperature monitoring; External (Ø×H): 36×18 mm
Catalog # ESPIDLDISK3618_CL
New Product
Supplier:   RAYPA
Temperature Data Loggers
Catalog # ESPIDLDISK3618_CL
Supplier:  RAYPA
New Product
Temperature Data Loggers
Temperature Data Loggers
AISI-316L stainless steel disk temperature recorder DLDISK3618_CL, 80 g, for accurate and precise temperature monitoring; External (Ø×H): 36×18 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
AISI-316L stainless steel disk temperature recorder with connection base and software.
  • User-friendly operation
  • Data logger available in multiple sizes to suit different needs
  • Delivers high accuracy and precision
Recommended for autoclave validation and for monitoring the internal temperature of containers.
Specifications
  • Description:
    AISI-316L stainless steel disk temperature recorder DLDISK3618_CL, 80 g, for accurate and precise temperature monitoring; External (Ø×H): 36×18 mm
  • Seq:
    2