Temperature Data Loggers
AISI-316L stainless steel disk temperature recorder with connection base and software BDL-DISK3618_CL, 200 g, for autoclave validation and for monitoring the internal temperature of containers, 60×95×175 mm (External)
Artikelnr. ESPIBDL-DISK3618_C
New Product
Anbieter: RAYPA
Jump to:
Product Details & Documents
AISI-316L stainless steel disk temperature recorder with connection base and software.
- User-friendly operation
- Data logger available in multiple sizes to suit different needs
- Delivers high accuracy and precision
Recommended for autoclave validation and for monitoring the internal temperature of containers.
Specifications
- Description:AISI-316L stainless steel disk temperature recorder with connection base and software BDL-DISK3618_CL, 200 g, for autoclave validation and for monitoring the internal temperature of containers
- Seq:3
- W×D×H:60×95×175 mm (External)