Orders LinkContactUsLinkComponent
Temperature Data Loggers
Temperature Data Loggers
Stainless steel and plastic connection base BDLDISK_CL, 120 g, for communication between the data logger and the software installed on a PC, 60×95×175 mm (External)
Catalog # ESPIBDLDISK_CL
New Product
Supplier:   RAYPA
Temperature Data Loggers
Catalog # ESPIBDLDISK_CL
Supplier:  RAYPA
New Product
Temperature Data Loggers
Temperature Data Loggers
Stainless steel and plastic connection base BDLDISK_CL, 120 g, for communication between the data logger and the software installed on a PC, 60×95×175 mm (External)
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
AISI-316L stainless steel disk temperature recorder with connection base and software.
  • User-friendly operation
  • Data logger available in multiple sizes to suit different needs
  • Delivers high accuracy and precision
Recommended for autoclave validation and for monitoring the internal temperature of containers.
Specifications
  • Description:
    Stainless steel and plastic connection base BDLDISK_CL, 120 g, for communication between the data logger and the software installed on a PC
  • Seq:
    1
  • W×D×H:
    60×95×175 mm (External)