Orders LinkContactUsLinkComponent
MPC Coupling Caps
MPC Coupling Caps
SaniQuik USP class VI, ADCF silicone seal, ¾", 100 pk, silicone, Silicone
Catalog # MFLX30003-00
New Product
MPC Coupling Caps
Catalog # MFLX30003-00
New Product
MPC Coupling Caps
MPC Coupling Caps
SaniQuik USP class VI, ADCF silicone seal, ¾", 100 pk, silicone, Silicone
Price
174,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
MPC connectors are multi-purpose, open-format connectors for use in a wide variety of critical fluid handling applications. This particular type of connector is perfect for smaller bag systems for aliquoted media or other products stored in bags.
  • Ergonomic thumb latch: Easy to operate - even with gloved hands
  • USP Class VI materials: Meet biocompatibility
  • Sterilisable by autoclave, EtO, e-beam, or gamma: Robust and economical for single-use applications
  • Parting line-free hose barb: Eliminates potential leak path
  • BSE/TSE free materials: Meet ADCF requirements
MPC connectors add ease of use and security to the most critical fluid handling applications. Choose from a full line of male and female MPC connectors and configurations, including pressure sealing caps and plugs, in sizes to fit ¹/₈", ¹/₄", and ³/₈" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
Specifications
  • Description:
    SaniQuik USP class VI, ADCF silicone seal, ¾", 100 pk, silicone
  • Seq:
    9
  • Fitting material:
    Silicone