Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Decoquinate 98%

New Product
Supplier:   Apollo Scientific
Decoquinate 98%
Supplier:  Apollo Scientific
New Product
Certificates

6 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester
  • CAS Number:18507-89-6
  • Formula:C₂₄H₃₅NO₅
  • Molecular Weight:417,54 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 155,00 €
    155,00 €
    1 * 100 g
       155,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 604,00 €
    604,00 €
    1 * 500 g
       604,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 2 950,00 €
    2 950,00 €
    1 * 2,5 kg
       2 950,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 43,50 €
    43,50 €
    1 * 25 g
       43,50 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product