Phenol Red ≥95%
Bottle
Catalog # APOSBUP19201-1G
New Product
Supplier: Apollo Scientific
Phenol Red ≥95%
Catalog # APOSBUP19201-1G
Supplier: Apollo Scientific
New Product
CAS Number: 143-74-8
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:143-74-8
- Formula:C₁₉H₁₄O₅S
- Molecular Weight:354,38 g/mol
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



