Order Entry
Austria
Orders LinkContactUsLinkComponent
Decoquinate ≥95%

Decoquinate ≥95%

New Product
Supplier:   Apollo Scientific
Decoquinate ≥95%
Supplier:  Apollo Scientific
New Product
Certificates
Decoquinate ≥95%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester
  • CAS Number:18507-89-6
  • Formula:C₂₄H₃₅NO₅
  • Molecular Weight:417,54 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 50,70 €
    50,70 €
    1 * 50 mg
       50,70 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 72,40 €
    72,40 €
    1 * 100 mg
       72,40 €

    Specifications:
    New Product
    New Product