Order Entry
Austria
Orders LinkContactUsLinkComponent
Fumaric acid 99%

Fumaric acid 99%

Irritant
New Product
Supplier:   Apollo Scientific
Fumaric acid 99%
Supplier:  Apollo Scientific
New Product
Certificates
Fumaric acid 99%

4 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116,07 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 21,80 €
    21,80 €
    1 * 100 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 57,90 €
    57,90 €
    1 * 500 g
       57,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 261,00 €
    261,00 €
    1 * 2,5 kg
       261,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 2 540,00 €
    2 540,00 €
    1 * 25 kg
       2 540,00 €

    Specifications:
    New Product
    New Product