CLASSIQSwabs™ Regular Polyester Swabs
Single polyester regular swab, with 100 mm breakpoint, Polyester, +
Catalog # COPA1U054S01
New Product
Supplier: Copan
Jump to:
Product Details & Documents
Traditional polyester nasal
Individually packaged
Sterile
Packaging: 100 swabs per box, 1000 swabs per case.
Specifications
- Sterile:+
- Description:Single polyester regular swab, with 100 mm breakpoint
- Material:Polyester