Order Entry
Austria
Orders LinkContactUsLinkComponent
cDNA Synthesis Kit | Avantor

cDNA Synthesis Kit

cDNA synthesis kit for miRNA, 50
Catalog # ORIGHP100042
New Product
Supplier:   OriGene
cDNA Synthesis Kit
Catalog # ORIGHP100042
Supplier:  OriGene
New Product
CAS Number:  
cDNA Synthesis Kit

cDNA synthesis kit for miRNA, 50

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents

qSTAR first-strand cDNA synthesis kit for miRNA is a two-step protocol kit.

The miRNA cDNA then becomes an effective qPCR template in SYBR Green I based qPCR method. The kit is uniquely designed to function only with OriGene’s miRNA detection system.

Delivery information: Kit includes 1 Vial poly (A) polymerase reaction 10X buffer [55 µl], 1 Vial poly (A) polymerase (5U/ μl) [55 µl], 1 Vial 10 mM ATP [55 µl], 1 Vial 5X MMLV buffer (dNTPs included)[350 µl], 1 Vial MMLV Reverse Transcriptase (200 U/µl)[55 µl], 1 Vial Oligo dT mix [55 µl], 1 Vial control primer pair (U6) [500 µl].

Specifications

Specifications

  • Description:
    cDNA synthesis kit for miRNA
  • No. of reactions:
    50
  • Cat. No.:
    ORIGHP100042
  • Storage conditions:
    Should be stored at –20 °C

Specifications

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.