Cellroll Set
New Product
Supplier: Binder
Jump to:
About this item
The Integra Cellroll is a compact and reliable rotary bottle system for cultivating adherent cells in roller bottles. This modular, expandable roller bottle system is suitable for either 4 or 6 roller bottles.
- Infinitely variable speed range from 0.1 to 2 rpm, the system enables gentle and uniform rotation of the cell culture bottles Ideal for large-volume cell cultures in research and production Continuous rotation ensures an even distribution of cells and culture medium, which promotes optimal growth conditions and high yields Specially designed for use in CO₂ incubators - with temperature-resistant materials and a compact, stackable design Fine adjustment of rotation direction, rotation speed: Fine adjustment of pause times and interval cycles: With soft start and stop phase:
2 Options Available Below
Product Details & Documents
The Integra Cellroll is a compact and reliable rotary bottle system for cultivating adherent cells in roller bottles. This modular, expandable roller bottle system is suitable for either 4 or 6 roller bottles.
- Infinitely variable speed range from 0.1 to 2 rpm, the system enables gentle and uniform rotation of the cell culture bottles Ideal for large-volume cell cultures in research and production Continuous rotation ensures an even distribution of cells and culture medium, which promotes optimal growth conditions and high yields Specially designed for use in CO₂ incubators - with temperature-resistant materials and a compact, stackable design Fine adjustment of rotation direction, rotation speed: Fine adjustment of pause times and interval cycles: With soft start and stop phase:
Product Family Options
Product Information
- AvailabilityPrice
- BIND8012-1885CELLROLL Set, 4 roller bottles for CB 170, CB 170-L, CB 170-O₂, CB 170-O₂L, CBF 170, CBF 170-O₂CELLROLL Set, 4 roller bottles for CB 170, CB 170-L, CB 170-O₂, CB 170-O₂L, CBF 170, CBF 170-O₂Log in for availability7 530,00 €Specifications:New ProductNew Product
- BIND8012-0572CELLROLL Set, 6 roller bottles for CB 260, CB 260-O₂, CBF 260, CBF 260-O₂CELLROLL Set, 6 roller bottles for CB 260, CB 260-O₂, CBF 260, CBF 260-O₂Log in for availability1 * 1 ST8 820,00 €Specifications:New ProductNew Product