EdgePLUS Digital Microscopes
About this item
The microscope is designed to provide high fidelity colour reproduction with its low-light performance and flexible LED control features for observing even your target’s most fine details.
High quality optics
1,3 megapixels
Flexible LED control (FLC)
Robust metallic housing
Adjustable polariser
The optics adopted in the Edge series reveals the finest details, answering the needs of the most demanding microscopy applications. The advanced CMOS image sensor can transmit fluid and crisp images at 30 FPS with resolution up to 1280×960.
Tasking with software, the FLC maximises illumination flexibility by offering independent on/off control of the four LED quadrants in addition to the 6-levels intensity adjustment capability. The robust metal housing made of anodised alloy protects the Dino-lite from UV and electromagnetic interferences.
The built-in adjustable polariser allows to remove freely the unwanted reflection or glare from the object surface for a better contrast.
6 Options Available Below
Product Details & Documents
The microscope is designed to provide high fidelity colour reproduction with its low-light performance and flexible LED control features for observing even your target’s most fine details.
High quality optics
1,3 megapixels
Flexible LED control (FLC)
Robust metallic housing
Adjustable polariser
The optics adopted in the Edge series reveals the finest details, answering the needs of the most demanding microscopy applications. The advanced CMOS image sensor can transmit fluid and crisp images at 30 FPS with resolution up to 1280×960.
Tasking with software, the FLC maximises illumination flexibility by offering independent on/off control of the four LED quadrants in addition to the 6-levels intensity adjustment capability. The robust metal housing made of anodised alloy protects the Dino-lite from UV and electromagnetic interferences.
The built-in adjustable polariser allows to remove freely the unwanted reflection or glare from the object surface for a better contrast.
Specifications for Product Family
Specifications
- Catalog No:
- IDCPAM4117MZT
- IDCPAM4517MZTL
- IDCPAM4517MZT
- IDCPAM4517MT-RFYW
- IDCPAM4917MT8
- IDCPAM4917MZT4
- Frame rate:
- 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
- 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
- 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
- 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
- 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
- 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
- Computer requirements:
- CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
- CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
- CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
- CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
- CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
- Windows 7, 8, 10, 11, MacOS 10.12 and up
- Software:
- DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
- DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
- DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
- DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
- DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
- DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
- Weight:
- 90 g
- 130 g
- 90 g
- 90 g
- 130 g
- 130 g
Specifications
Product Family Options
Product Information
- ModelMagnificationIlluminationResolutionAvailabilityPrice
- IDCPAM4117MZTDigital microscope, Ø: 33 mm, Height: 104 mmDigital microscope, Ø: 33 mm, Height: 104 mmAM4117MZT10 - 220XWhite light, 8 LEDs1,3 Megapixel (1280×960)Log in for availability1 * 1 ST797,00 €Specifications:New Product
More
Specifications
Cat. No.IDCPAM4117MZTFrame rate30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEGComputer requirementsCPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or moreSoftwareDinoCapture 2.0 (Windows), DinoXcope (Mac OS)Weight90 gNew Product - IDCPAM4517MZTLDigital microscope, Ø: 33 mm, Length: 105 mmDigital microscope, Ø: 33 mm, Length: 105 mmAM4517MZTL10 - 140XWhite light, 8 LEDs1,3 Megapixel (1280×960)Log in for availability1 * 1 ST857,00 €Specifications:New Product
More
Specifications
Cat. No.IDCPAM4517MZTLFrame rate30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEGComputer requirementsCPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or moreSoftwareDinoCapture 2.0 (Windows), DinoXcope (Mac OS)Weight130 gNew Product - IDCPAM4517MZTDigital microscope, Ø: 33 mm, Length: 105 mmDigital microscope, Ø: 33 mm, Length: 105 mmAM4517MZT10 - 220XWhite light, 8 LEDs1,3 Megapixel (1280×960)Log in for availability1 * 1 ST857,00 €Specifications:New Product
More
Specifications
Cat. No.IDCPAM4517MZTFrame rate30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEGComputer requirementsCPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or moreSoftwareDinoCapture 2.0 (Windows), DinoXcope (Mac OS)Weight90 gNew Product - IDCPAM4517MT-RFYWDigital microscope, Ø: 32 mm, Height: 104 mmDigital microscope, Ø: 32 mm, Height: 104 mmAM4517MT-RFYW20 - 220XWhite light, 8 LEDs1,3 Megapixel (1280×960)Log in for availability1 * 1 ST985,00 €Specifications:New Product
More
Specifications
Cat. No.IDCPAM4517MT-RFYWFrame rate30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEGComputer requirementsCPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or moreSoftwareDinoCapture 2.0 (Windows), DinoXcope (Mac OS)Weight90 gNew Product - IDCPAM4917MT8Digital microscope, Ø: 33 mm, Height: 107 mmDigital microscope, Ø: 33 mm, Height: 107 mmAM4917MT8700 - 900XWhite light, 8 LEDs1,3 Megapixel (1280×960)Log in for availability1 * 1 ST1 120,00 €Specifications:New Product
More
Specifications
Cat. No.IDCPAM4917MT8Frame rate30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEGComputer requirementsCPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or moreSoftwareDinoCapture 2.0 (Windows), DinoXcope (Mac OS)Weight130 gNew Product - IDCPAM4917MZT4Digital microscope, Ø: 33 mm, Height: 107 mmDigital microscope, Ø: 33 mm, Height: 107 mmAM4917MZT4400 ~ 470xWhite light, 8 LEDs1,3 Megapixel (1280×960)Log in for availability1 * 1 ST1 120,00 €Specifications:New Product
More
Specifications
Cat. No.IDCPAM4917MZT4Frame rate30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEGComputer requirementsWindows 7, 8, 10, 11, MacOS 10.12 and upSoftwareDinoCapture 2.0 (Windows), DinoXcope (Mac OS)Weight130 gNew Product