Orders LinkContactUsLinkComponent
Dispensing Lines
Dispensing Lines
New Product
Supplier:   RAYPA
Dispensing Lines
Supplier:  RAYPA
New Product
Certificates
Dispensing Lines
Dispensing Lines
3 Options Available Below
Product Details & Documents
Additional silicone dosing tubes, with metal nozzle for dispensing sterile culture media.
  • Silicone dosing tubes with press-fit connection and metal nozzle
  • Sterile medium is dispensed through the flame-sterilizable nozzle
  • Made of MVQ silicone; heat-resistant up to 200 °C; 60 A hardness
  • Available in multiple diameters
  • Steam and pressurised air enable easy line cleaning before, during, and after use
  • All models are autoclavable, but autoclaving after each use is unnecessary thanks to the built-in steam cleaning program
Ordering information: Dispensing lines can be customised, adjusting their length or adding modifications to adapt them to the specific production requirements of the customer.
Product Family Options
Product Information
  • Availability
    Price
  • ESPITUB-DOSIF-3.2
    Additional sets of dispensing lines, TUB-DOSIF-3,2, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 3,2 mm int.Ø, 6,4 mm ext.Ø
    Additional sets of dispensing lines, TUB-DOSIF-3,2, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 3,2 mm int.Ø, 6,4 mm ext.Ø
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • ESPITUB-DOSIF-4
    Additional sets of dispensing lines, TUB-DOSIF-4, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 4 mm int.Ø, 7,2 mm ext.Ø
    Additional sets of dispensing lines, TUB-DOSIF-4, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 4 mm int.Ø, 7,2 mm ext.Ø
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product
  • ESPITUB-DOSIF-8
    Additional sets of dispensing lines TUB-DOSIF-8, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 8 mm int.Ø, 11,2 mm ext.Ø
    Additional sets of dispensing lines TUB-DOSIF-8, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 8 mm int.Ø, 11,2 mm ext.Ø
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    New Product
    New Product