Software SWMP
Communication software between the equipment and the PC, SWMP, The visualisation and printing of the processes carried out
Catalog # ESPISWMP
New Product
Supplier: RAYPA
Jump to:
Product Details & Documents
Communication software between the equipment and the PC that allows the display and recording in real time or after each cycle performed in the media preparator.
Included items: ethernet cable 1,8 m in length, ethernet to us adapter and USB memory stick with the software
- Allows in-depth analysis of data related to each cycle
- Intuitive display of information on the cycles performed: batch record, outcome, probes temperature, pressure, serial number, autoclave model, errors, etc
- Access is protected by username and password
- Available languages are: English, Spanish, French and Italian
- The export of memory cycles is encrypted (.DAT files)
Specifications
- Description:Communication software between the equipment and the PC, SWMP
- For:The visualisation and printing of the processes carried out
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.