Label Printer
Direct thermal label printer for Printing individual labels with barcodes and identifying data for each processed load, Resolution: 203 dpi, Printing individual labels with barcodes and identifying data for each processed load
Catalog # ESPIITS-LAB
Supplier: RAYPA
Jump to:
Product Details & Documents
Allows printing individual labels with barcodes and identifying data for each processed load.
The purchase of this accessory includes: 1 label printer, 1 power adapter, 1 power cable, 1 communication cable and 1 roll of label paper
- Connection: RS232
All top line autoclaves include RAYPAtrace, a simple and professional solution for the documentation and complete traceability of processed batches. To optimise the process, it is recommended to use a label printer (Ref. ITS-LAB) and a barcode scanner (Ref. BAR-SCAN), tools that facilitate the identification and tracking of each batch through individual labels.
Specifications
- Description:Direct thermal label printer for Printing individual labels with barcodes and identifying data for each processed load, Resolution: 203 dpi
- For:Printing individual labels with barcodes and identifying data for each processed load