Orders LinkContactUsLinkComponent
Filter Vial Toggle Press
Filter Vial Toggle Press
Filter vial toggle press
Catalog # THMS35005
Supplier:   THOMSON INSTRUMENT
Filter Vial Toggle Press
Catalog # THMS35005
Supplier:  THOMSON INSTRUMENT
Filter Vial Toggle Press
Filter Vial Toggle Press
Filter vial toggle press
Price
571,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The Thomson filter vial toggle press quickly depresses autosampler-ready filter vials.
  • Filter vial toggle press depresses five filter vials at one time
Specifications
  • Description:
    Filter vial toggle press