Order Entry
Austria
Orders LinkContactUsLinkComponent
D-(+)-Galactose ≥98% C-4 epimer of glucose

D-(+)-Galactose ≥98% C-4 epimer of glucose

Catalog # ABCAAB146237-25
Marketsource
Supplier:   Abcam
D-(+)-Galactose ≥98% C-4 epimer of glucose
Catalog # ABCAAB146237-25
Supplier:  Abcam
Marketsource
CAS Number:  59-23-4
D-(+)-Galactose ≥98% C-4 epimer of glucose
CAS Number:  59-23-4
Price
76,90 €
Log in for availability
1 * 25 g
Product Details & Documents

C-4 epimer of glucose. Aldohexose that serves as an energy source and as an essential component of glycolipids and glycoproteins.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Molecular Weight:180,16 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% D-(+)-Galactose