Order Entry
Austria
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid ≥98% antioxidant

L(+)-Ascorbic acid ≥98% antioxidant

Catalog # ABCAAB142830-100
Marketsource
Supplier:   Abcam
L(+)-Ascorbic acid ≥98% antioxidant
Catalog # ABCAAB142830-100
Supplier:  Abcam
Marketsource
CAS Number:  50-81-7
L(+)-Ascorbic acid ≥98% antioxidant
CAS Number:  50-81-7
Price
118,00 €
Log in for availability
1 * 100 g
Product Details & Documents

L-(+)-Ascorbic acid, antioxidant. Achieve your results faster with highly validated, pure and trusted compounds.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Molecular Weight:176,12 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Ascorbic acid