Order Entry
Austria
Orders LinkContactUsLinkComponent
Western Blot Stripping Buffer, Abcam

Western Blot Stripping Buffer, Abcam

Western Blot Stripping Buffer
Catalog # ABCAAB282569-500
Marketsource
Supplier:   Abcam
Western Blot Stripping Buffer, Abcam
Catalog # ABCAAB282569-500
Supplier:  Abcam
Marketsource
CAS Number:  
Western Blot Stripping Buffer, Abcam

Western Blot Stripping Buffer

CAS Number:  
Price
262,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Western Blot Stripping Buffer (ab282569) uniformly removes antibodies from Western blots so that the blots may be reprobed with different antibodies.

  • pH: 2.9 - 3.1 Constituents: 88% Water, 10% Sodium lauryl sulfate

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Western Blot Stripping Buffer
  • Cat. No.:
    ABCAAB282569-500
  • Storage temperature:
    Store at Ambient.

Specifications