Order Entry
Austria
Orders LinkContactUsLinkComponent
Extraction Buffer 5X PTR, Abcam

Extraction Buffer 5X PTR, Abcam

Marketsource
Supplier:   Abcam
Extraction Buffer 5X PTR, Abcam
Supplier:  Abcam
Marketsource
Certificates

2 Options Available Below

Product Details & Documents

Extraction buffer 5X PTR is formulated for preparing cell or tissue extracts for use in immunoassays, with added phosphatase inhibitors and protease inhibitor aprotinin. - Cited in over 20 publications - Phosphatase and protease inhibitors – contains inhibitors to preserve sample integrity - Easy to use – dilute to 1X with distilled water before use, or use with extraction enhancer buffer 50X for enhanced extraction - Compatible with a variety of sample types

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCAAB193970-10
  • ABCAAB193970-50
  • Storage temperature:
  • Store at +4°C.
  • Store at +4°C.

Specifications

Product Family Options

unology General

Product Information

  • Availability
    Price
  • ABCAAB193970-10
    Extraction Buffer 5X PTR
    Extraction Buffer 5X PTR
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - 56,40 €
    56,40 €
    1 * 10 mL
       56,40 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB193970-10
    Storage temperature
    Store at +4°C.
    Marketsource

Product Information

  • Availability
    Price
  • ABCAAB193970-50
    Extraction Buffer 5X PTR
    Extraction Buffer 5X PTR
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 71,80 €
    71,80 €
    1 * 50 mL
       71,80 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB193970-50
    Storage temperature
    Store at +4°C.
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.