Order Entry
Austria
Orders LinkContactUsLinkComponent
ChIP Buffer, Abcam

ChIP Buffer, Abcam

ChIP Buffer
Catalog # ABCAAB205698-6
Marketsource
Supplier:   Abcam
ChIP Buffer, Abcam
Catalog # ABCAAB205698-6
Supplier:  Abcam
Marketsource
CAS Number:  
ChIP Buffer, Abcam

ChIP Buffer

CAS Number:  
Price
134,00 €
Log in for availability
1 * 6 mL
Product Details & Documents

ChIP Buffer for ab185913

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    ChIP Buffer
  • Cat. No.:
    ABCAAB205698-6
  • Storage temperature:
    Store at +4°C.

Specifications