Orders LinkContactUsLinkComponent
10X Blocking Buffer, Abcam
10X Blocking Buffer, Abcam
10X Blocking Buffer
Catalog # ABCAAB126587-50
Marketsource
Supplier:   Abcam
10X Blocking Buffer, Abcam
Catalog # ABCAAB126587-50
Supplier:  Abcam
Marketsource
CAS Number:  
10X Blocking Buffer, Abcam
10X Blocking Buffer
CAS Number:  
Price
82,00 €
Log in for availability
1 * 50 mL
Product Details & Documents
10X blocking buffer (pH 7.5) for serum-free blocking of non-specific antibody binding. - Cited in over 30 publications - Suitable for ELISA, in-cell ELISA, ICC/IF, western blot (WB) - 100% casein blocking as preservative
Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
  • Description:
    10X Blocking Buffer