Order Entry
Austria
ContactUsLinkComponent
Scan® 5000 AI Automatic AI Colony Counter and Inhibition Zone Reader
Scan® 5000 AI Automatic AI Colony Counter and Inhibition Zone Reader
Catalog # INSC434500
Supplier:  INTERSCIENCE
New Product
Scan® 5000 AI Automatic AI Colony Counter and Inhibition Zone Reader
Catalog # INSC434500
Supplier:  INTERSCIENCE
New Product

Specifications

  • Model:
    Scan 5000 AI
  • Resolution:
    12,2 megapixels
  • Description:
    Automatic AI colony counter and inhibition zone reader
  • Camera:
    Ultra HD CMOS (colour)
  • Count:
    Up to 1000 colonies per second
  • Dimensions:
    46,4×46,4×63,1 cm
  • Dish size:
    Ø 55 - 150 mm round Petri dishes and 120 mm square Petri dishes
  • Weight:
    25,5 kg

Specifications

About this item

Scan® 5000 Ai automates and standardizes colony counting for microbiological analyses. The power of Artificial Intelligence offers unparalleled performance and continuous development opportunities at the cutting edge of technological developments. It is suitable for all sizes of Petri dishes up to 150 mm and all culture media.

  • Minimum size of detected colony 0,03 mm

  • Counting on Ø 55 to 150 mm round Petri dishes and 120 mm square Petri dishes
  • Counting on pour, surface, Spiral, circle mode plated dishes
  • Counting on chromogenic agar, Petrifilm™, MC-media Pads™, Compact Dry™, Easy Plate™, filtration membranes
  • Inhibition zone reader
  • AI-powered colony counting

Unrivaled precision with artificial intelligence. The power of artificial intelligence of the Scan® Ai automatic counter boosts the analysis of Petri dishes. Scan® Ai standardizes colony counting and improves the accuracy of your analysis results. The scientific expertise of the technical teams remains within the lab.

White LED dome lighting, reflection and shadow free Petri dishes are difficult to light, because they are transparent and reflective. Heterogeneous lighting often creates artifacts on the meniscus of the agar and on the edge of the plate. These artifacts are counted as colonies or prevent perfect counting.

We have designed a white diffusing dome that provides 360° lighting for the plate, without any reflection or shadow. The light is diffused from all points, and provides lighting without artifacts for counting even on the edge of the plate.

USB data export: Recountable session, Excel™, PDF report, jpg, png and bmp images
Data security: Modified data traceability in conformity with 21 CFR part 11

Certifications:

CE. UKCA. ISO 9001. RoHS. WEEE.

Accessories information:

Perpetual license Scan® 5000 AI. License for automatic AI colony counter and inhibition zone reader.

The license of Scan® 5000 Ai automates and standardizes colony counting for microbiological analyses. The power of Artificial Intelligence offers unparalleled performance.

Software: user-friendly and easy-to-use. Live image on the software. Instant result. Data integrity: electronic signature, audit trail. Account managment. Multiple exports data possibilities.

Data integrity. Scan® Ai receives and sends information from LIMS and/or dataLink and digitizes the counting. All the data from the analysis are traceable and auditable. The results are validated by a double electronic signature. Thanks to account management, you have multi-level access for the validation of results. This solution thus ensures the reliability and security of data in accordance with FDA guidelines, 21 CFR Part 11 and GMP (Good Manufacturing Practices) Annex 11.

Traceability and printing of results. Scan® Ai connects to the LIMS in bi-directionality and can be connected to the diluter and plater via dataLink, a traceability system. It offers multiple export possibilities to save time on manual entry and thus increase the security and quality of your analyses.

Export your results to your PC or archive them in CSV, Excel™, OpenOffice™, PDF, SCA, BIO format. You can also export images from the camera in JPEG, PNG and BMP format.

Delivery: Key license number.

Ordering information:

The license (accessory) is sold separately.

Delivery:

Supplied with 1×15 V power supply, 1×USB cable, 3 validation plates, 1×user manual, 1×conformity certificate/warranty card.