Order Entry
Austria
ContactUsLinkComponent
Scan® 3000 AI Automatic AI Colony Counter and Inhibition Zone Reader
Scan® 3000 AI Automatic AI Colony Counter and Inhibition Zone Reader
Catalog # INSC434300
Supplier:  INTERSCIENCE
New Product
undefined
Scan® 3000 AI Automatic AI Colony Counter and Inhibition Zone Reader
Catalog # INSC434300
Supplier:  INTERSCIENCE
New Product

Specifications

  • Model:
    Scan® 3000 AI
  • Resolution:
    5 megapixels
  • Description:
    Automatic AI colony counter and inhibition zone reader
  • Camera:
    Ultra HD CMOS (colour)
  • Count:
    Up to 1000 colonies per second
  • Dimensions:
    32×32×44 cm
  • Dish size:
    Ø 55 - 90 mm (Petri)
  • Weight:
    11,5 kg

Specifications

About this item

Scan 3000 Ai is a cutting-edge tool designed to automate and standardize colony counting in microbiological analyses. Thanks to artificial intelligence, it offers exceptional performance and is constantly evolving to incorporate the latest technological advances. It is compatible with all sizes of Petri dishes up to 90 mm and all types of culture media.

  • Minimum size of detected colony is 0,03 mm

  • Counting on round Petri dishes with a diameter of 55 to 90 mm
  • Counting on pour, surface, Spiral and circle plating
  • Counting on chromogenic agars, Petrifilm™, MC-media Pads™, Compact Dry™, Easy Plate™, filtration membranes
  • Reading of inhibition zones
  • AI-powered colony counting

Unrivaled precision with artificial intelligence. The power of artificial intelligence of the Scan Ai automatic counter boosts the analysis of Petri dishes. Scan Ai standardizes colony counting and improves the accuracy of your analysis results. The scientific expertise of the technical teams remains within the lab.

White LED dome lighting, reflection and shadow free. Petri dishes are difficult to light, because they are transparent and reflective. Heterogeneous lighting often creates artifacts on the meniscus of the agar and on the edge of the plate. These artifacts are counted as colonies or prevent perfect counting.

We have designed a white diffusing dome that provides 360° lighting for the plate, without any reflection or shadow. The light is diffused from all points, and provides lighting without artifacts for counting even on the edge of the plate.

USB data export: Recountable session, Excel™, PDF report, jpg, png and bmp images
Data security: Modified data traceability in conformity with 21 CFR part 11

Certifications:

CE. UKCA. ISO 9001. RoHS. WEEE.

Accessories information:

The license of Scan 3000 Ai is a cutting-edge tool designed to automate and standardize colony counting in microbiological analyses. Thanks to artificial intelligence, it offers exceptional performance.

Software: user-friendly and easy-to-use. Live image on the software. Instant result. Data integrity: electronic signature, audit trail. Account managment. Multiple exports data possibilities.

Data integrity. Scan Ai receives and sends information from LIMS and/or dataLink and digitizes the counting. All the data from the analysis are traceable and auditable. The results are validated by a double electronic signature. Thanks to account management, you have multi-level access for the validation of results. This solution thus ensures the reliability and security of data in accordance with FDA guidelines, 21 CFR Part 11 and GMP (Good Manufacturing Practices) Annex 11.

Traceability and printing of results. Scan Ai connects to the LIMS in bi-directionality and can be connected to the diluter and plater via dataLink, a traceability system. It offers multiple export possibilities to save time on manual entry and thus increase the security and quality of your analyses.

Export your results to your PC or archive them in CSV, Excel™, OpenOffice™, PDF, SCA, BIO format. You can also export images from the camera in JPEG, PNG and BMP format.

Delivery: Key license number

Ordering information:

Need to be ordered. The license (accessory) is sold separately.

Delivery:

Supplied with 1×15 V power supply, 1×USB cable, 3×validation plates, 1×user manual, 1×conformity certificate/warranty card.