Order Entry
Austria
ContactUsLinkComponent
Proteometer-CV Fluorescent Reagent
Proteometer-CV Fluorescent Reagent
Catalog # NOVIRGN-PCV-001
Supplier:  NOVILYTIC
New Product
CAS Number:  
undefined
Proteometer-CV Fluorescent Reagent
Catalog # NOVIRGN-PCV-001
Supplier:  NOVILYTIC
New Product
CAS Number:  

Specifications

  • Description:
    Proteometer-CV fluorescent reagent. 1 vial of fluorescent reagent for the Proteometer-CV kit

Specifications

About this item

Replacement fluorescent kit reagent for the Proteometer-CV.

  • Replacement vials for frequent users of the Proteometer-CV kit
  • Store at −20 °C

Intended for use with the Proteometer-CV kit. Crimped, sealed, sold individually.

Packaging: Supplied in a custom box with instructions.