Proteometer-CV Reactor
Catalog # NOVIRCT-PCV-001
Supplier: NOVILYTIC
New Product
Specifications
- Description:Reactor for Proteometer-CV kit
Specifications
About this item
Reactor for Proteometer-CV kit.
- Plugs into any LC with a fluorescence detector
- Store at room temperature
The Proteometer-CV is designed for the scientific development of mAbs, bi-specifics, or fusion proteins with a human Fc region, regardless of their stage in the development continuum.
Packaging: Supplied in a custom box with instructions.