Order Entry
Austria
ContactUsLinkComponent
Proteometer-CV Reconstitution Reagent
Proteometer-CV Reconstitution Reagent
Catalog # NOVIREC-PCV-001
Supplier:  NOVILYTIC
New Product
CAS Number:  
undefined
Proteometer-CV Reconstitution Reagent
Catalog # NOVIREC-PCV-001
Supplier:  NOVILYTIC
New Product
CAS Number:  

Specifications

  • Description:
    Proteometer-CV reconstitution reagent, 1×5 ml vial of reconstitution reagent for the Proteometer-CV kit

Specifications

About this item

Proteometer-CV reconstitution reagent.

  • Proteometer-CV reconstitution reagent
  • Store at room temperature

Intended for use with the Proteometer-CV kit. Reorder individually as needed.

Packaging: Supplied in a custom box with instructions.