ContactUsLinkComponent
Brady® BBP®12 Printer Kit with Vial Labels
Brady® BBP®12 Printer Kit with Vial Labels
Supplier:  Brady
New Product
Certificates
undefined
Brady® BBP®12 Printer Kit with Vial Labels
Supplier:  Brady
New Product
Specifications
  • Catalog No:
  • BRDY324280
  • BRDY324279
  • Connections:
  • Ethernet; Serial; USB
  • Ethernet; Serial; USB
  • Model:
  • BBP®12
  • BBP®12
  • Power supply:
  • EU plug
  • UK plug
  • Print width:
  • 105,41 mm (max.)
  • 105,41 mm (max.)
  • Width:
  • 116,84 mm (max, label/tape)
  • 116,84 mm (max, label/tape)