ContactUsLinkComponent
Brady® M510 Printer Kit with Slide Labels
Brady® M510 Printer Kit with Slide Labels
Supplier:  Brady
New Product
Certificates
undefined
Brady® M510 Printer Kit with Slide Labels
Supplier:  Brady
New Product
Specifications
  • Catalog No:
  • BRDY324276
  • BRDY324275
  • Connections:
  • USB
  • USB
  • Model:
  • M510
  • M510
  • Power supply:
  • EU plug
  • UK plug
  • Print width:
  • 36,58 mm (max.)
  • 36,58 mm (max.)
  • Width:
  • 38,10 mm (max, label/tape)
  • 38,10 mm (max, label/tape)