Order Entry
Austria
ContactUsLinkComponent
Proteometer-UFT Buffer Packets
Catalog # NOVIBUF-PUF-001
Supplier:  NOVILYTIC
New Product
CAS Number:  
undefined
Proteometer-UFT Buffer Packets
Catalog # NOVIBUF-PUF-001
Supplier:  NOVILYTIC
New Product
CAS Number:  

Specifications

  • Description:
    Proteometer-UFT buffer packets

Specifications

About this item

Specifically formulated buffer packets for use with the Proteometer-UFT kit, sold in sets of 3.

  • Replacement buffer packets allow users to reorder when reactor and/or reagents are not required
  • Store at room temperature

Intended for use with the Proteometer-UFT kit. Reorder individually as needed.

Ordering information: No refrigeration or special handling required.
Packaging: Supplied in a custom box with instructions.