Orders LinkContactUsLinkComponent
Proteometer-UFT MD unit
Proteometer-UFT MD unit
Proteometer-UFT MD unit
Catalog # NOVIRCT-PUF-001
New Product
Supplier:   NOVILYTIC
Proteometer-UFT MD unit
Catalog # NOVIRCT-PUF-001
Supplier:  NOVILYTIC
New Product
Proteometer-UFT MD unit
Proteometer-UFT MD unit
Proteometer-UFT MD unit
Price
704,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Proteometer-UFT MD unit sold individually.
  • Proteometer-UFT MD unit replaces the LC column in the analytical system
Intended for use with the Proteometer-UFT kit.
 : Reorder individually as needed.
 : Supplied in a custom box with instructions for use. Follow instructions in "Instructions for Use" for proper storage.
Specifications
  • Description:
    Proteometer-UFT MD unit