Order Entry
Austria
Orders LinkContactUsLinkComponent
Proteometer-UFT starter kit

Proteometer-UFT starter kit

Proteometer-UFT starter kit, reagent to be stored at -20 °C. For use with any LC equipped with a fluorescence detector
Catalog # NOVIKIT-PUF-002-01
New Product
Supplier:   NOVILYTIC
Proteometer-UFT starter kit
Catalog # NOVIKIT-PUF-002-01
Supplier:  NOVILYTIC
New Product
CAS Number:  
Proteometer-UFT starter kit
Proteometer-UFT starter kit

Proteometer-UFT starter kit, reagent to be stored at -20 °C. For use with any LC equipped with a fluorescence detector

CAS Number:  
Price
1 630,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

The Proteometer-UFT Kit transforms any LC into an analytical system capable of identifying and quantifying titer content in crude culture filtrate in <90 seconds, without sample preparation.

  • No system regeneration: Reduces analysis time & cost

  • No Protein A required: Hours to 90secs
  • Analysis of CFB samples: HCP removal no needed
  • Plugs into any LC: Easy adoption - no new software needed
  • No method development: Reduces time until method usage
  • No LC column required: Faster processing, long life
  • Adjustable titer range: One test for upstream & downstream
  • Low sample volume (≤ 20μL): Reduced impact on processing
  • At-line analysis: Reduces time between collection & analysis

Determination of titer of therapeutic mAbs and other proteins that contain the Fc domain of human IgG. It is intended primarily to monitor mAb concentration in growth media during clone selection and product development utilizing HPLC systems equipped with a fluorescence detector.

Accessories information: See the Proteometer-UFT MD Unit, Proteometer-UFT Buffer Packets, and Proteometer-UFT Reagent series for accessory component replacement options.

Delivery information: Includes 1x MD unit, 1x Proteometer-UFT Fluorescent reagent vial, 1x mixing tee, 2x tubing unions, 1x Proteometer-UFT buffer packet, 1x reconstitution reagent. Allows for analysis up to 300 samples.

Packaging: Supplied in a custom box with instructions for use.

Specifications

Specifications

  • Description:
    Proteometer-UFT starter kit, reagent to be stored at -20 °C. For use with any LC equipped with a fluorescence detector

Specifications