Proteometer-L Reactor
NOVIRCT-PL-001
New Product
- Description:Proteometer-L reactor
Replacement reactor for Proteometer-L kit.
- Plugs into any LC with a fluorescence detector
- Store at room temperature
Intended for use with the Proteometer-L kit. Reorder individually as needed.
: Supplied in a custom box with instructions.