ContactUsLinkComponent
FARRAR® ULC Ultra-Low Chambers
FARRAR® ULC Ultra-Low Chambers
Supplier:  TRANE TECHNOLOGIES
New Product
Certificates
FARRAR® ULC Ultra-Low Chambers
Supplier:  TRANE TECHNOLOGIES
New Product
Specifications
  • Catalog No:
  • FARRULC190-SAC50HZ
  • FARRULC259-SWC50HZ
  • FARRULC259-BWC50HZ
  • FARRULC328-SAC50HZ
  • FARRULC259-BAC50HZ
  • FARRULC259-SAC50HZ
  • FARRULC190-SWC50HZ
  • FARRULC328-SWC50HZ
  • FARRULC259-DAC50HZ
  • FARRULC259-DWC50HZ
  • Air circulation:
  • Forced air circulation
  • Forced air circulation
  • Forced air circulation
  • Forced air circulation
  • Forced air circulation
  • Forced air circulation
  • Forced air circulation
  • Forced air circulation
  • Forced air circulation
  • Forced air circulation
  • Display:
  • HMI touchscreen provides visual temperature profile progress with export capability
  • HMI touchscreen provides visual temperature profile progress with export capability
  • HMI touchscreen provides visual temperature profile progress with export capability
  • HMI touchscreen provides visual temperature profile progress with export capability
  • HMI touchscreen provides visual temperature profile progress with export capability
  • HMI touchscreen provides visual temperature profile progress with export capability
  • HMI touchscreen provides visual temperature profile progress with export capability
  • HMI touchscreen provides visual temperature profile progress with export capability
  • HMI touchscreen provides visual temperature profile progress with export capability
  • HMI touchscreen provides visual temperature profile progress with export capability
  • Defrost:
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Electrical:
  • 208-230 VAC, Single Phase, 50 Hz
  • 208-230 VAC, Single Phase, 50 Hz
  • 208-230 VAC, Single Phase, 50 Hz
  • 208-230 VAC, Single Phase, 50 Hz
  • 208-230 VAC, Single Phase, 50 Hz
  • 208-230 VAC, Single Phase, 50 Hz
  • 208-230 VAC, Single Phase, 50 Hz
  • 208-230 VAC, Single Phase, 50 Hz
  • 208-230 VAC, Single Phase, 50 Hz
  • 208-230 VAC, Single Phase, 50 Hz
  • Shelves/drawers:
  • Includes ULC-190 Standard Shelf Kit (3 shelves behind each door)
  • Includes ULC-259 Standard Shelf System (3 shelves per unit)
  • Includes ULC-259 Biorack Assembly. Designed to hold 112 of our 572 Cryoracks (accessory).
  • Includes ULC-328 Pallet Rack. Designed to accommodate 2 pallets for bulk storage behind each door.
  • Includes ULC-259 Biorack Assembly. Designed to hold 112 of our 572 Cryoracks (accessory).
  • Includes ULC-259 Standard Shelf System (3 shelves per unit)
  • Includes ULC-190 Standard Shelf Kit (3 shelves behind each door)
  • Includes ULC-328 Pallet Rack. Designed to accommodate 2 pallets for bulk storage behind each door.
  • Includes ULC-259 Drawer System which provides 6 drawers on each side of unit. Each drawer has 4 individual pans.
  • Includes ULC-259 Drawer System which provides 6 drawers on each side of unit. Each drawer has 4 individual pans.
  • Weight:
  • 1429 kg
  • 1588 kg
  • 1588 kg
  • 1769 kg
  • 1588 kg
  • 1588 kg
  • 1429 kg
  • 1769 kg
  • 1588 kg
  • 1588 kg
  • Shipping weight:
  • 1769 kg
  • 1860 kg
  • 1860 kg
  • 2132 kg
  • 1860 kg
  • 1860 kg
  • 1769 kg
  • 2132 kg
  • 1860 kg
  • 1860 kg