Order Entry
Austria
ContactUsLinkComponent
 
Chest Freezers, −25 to −45 °C, LT C
 :  TENAK
New Product
 
undefined
Chest Freezers, −25 to −45 °C, LT C
 :  TENAK
New Product

 

  • Catalog No:
  • TENAN111006
  • TENAN111002
  • TENAN111003
  • TENAN111005
  • TENAN111004
  • Temperature range:
  • −25...−45 °C
  • −25...−45 °C
  • −25...−45 °C
  • −25...−45 °C
  • −25...−45 °C
  • Model:
  • LT C200
  • LT C150
  • LT C300
  • LT C500
  • LT C400
  • Refrigerant:
  • R290
  • R290
  • R290
  • R290
  • R290
  • W×D×H ext.:
  • 923×680×885 mm
  • 725×680×855 mm
  • 1265×680×885 mm
  • 1664×740×885 mm
  • 1560×680×885 mm

 

Frequently Bought Together