ContactUsLinkComponent
Ultra Low Temperature Freezers
Ultra Low Temperature Freezers
Supplier:  TENAK
New Product
Certificates
Ultra Low Temperature Freezers
Supplier:  TENAK
New Product
Specifications
  • Catalog No:
  • TENAN123000
  • TENAN123001
  • Temperature range:
  • −60...−86 °C
  • −60...−86 °C
  • Model:
  • ULT U35
  • ULT U100
  • Refrigerant:
  • Nature R
  • Nature R
  • W×D×H ext.:
  • 630×675×650 mm
  • 595×630×825 mm
Specifications
Racks for ULT/XLT U100
Product Information
Frequently Bought Together