ContactUsLinkComponent
Fixed Angle Rotors for MIKRO 2.0 Centrifuges
Fixed Angle Rotors for MIKRO 2.0 Centrifuges
Supplier:  Hettich
   Sustainable Options Available
New Product
Certificates
undefined
Fixed Angle Rotors for MIKRO 2.0 Centrifuges
Supplier:  Hettich
   Sustainable Options Available
New Product
Specifications
  • Catalog No:
  • 521-4355
  • 521-4357
  • 521-4358
  • 521-4354
  • 521-4356
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Capacity:
  • 24 spin columns
  • 20 cryotubes
  • 12×5 ml microtubes
  • 24×2.0 ml
  • 8×8 PCR strips
  • Angle:
  • 45°
  • 45°
  • 45°
  • 45°
  • 45°
  • Max. RPM:
  • 14600
  • 14600
  • 14900
  • 16100
  • 15400
  • Environmental certification:
  • ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)