Order Entry
Austria
ContactUsLinkComponent
 
VWR® PFAS Kits, 0,7 ml PP Vials with 11 mm Snap Ring Cap, Silicone/Polypropylene
 
undefined
VWR® PFAS Kits, 0,7 ml PP Vials with 11 mm Snap Ring Cap, Silicone/Polypropylene
 

 

  • Catalog No:
  • 548-7007A
  • 548-7004A
  • Kit:
  • KIT
  • KIT
  • Component vial:
  • 548-7013A (kits 548-7004A, 548-7007A)
  • 548-7013A (kits 548-7004A, 548-7007A)
  • Component cap:
  • 548-7010A (kit 548-7004A); 548-7006A (kit 548-7007A)
  • 548-7010A (kit 548-7004A); 548-7006A (kit 548-7007A)
  • Height with cap:
  • 6,1 mm
  • 6,1 mm
  • Ext.Ø with cap:
  • 12 mm
  • 12 mm

 

Frequently Bought Together