Axygen® RIGIPLATE® PCR Microplates for Automation
Supplier: Corning
New Product
Certificates
About this item
These PP/PC PCR plates have maximum volumes that range from 40 to 330 µl and are compatible with most thermal cyclers.
ANSI/SBS foot print
- Smooth, thin, uniform well walls ensure accurate thermal transfer
- Wells are slightly raised to accommodate sealing mats, films or foils
- Plates are skirted to allow barcoding, and include a frosted labelling area
- Lot tested and certified free from DNase, RNase and human genomic DNA
- Printed alphanumeric labelling simplifies plate and sample identification
Ideal for high throughput screening, sample preparation and NGA library preparation.
Barcoded: No
Certifications:
DNase/RNAse-free, Human DNA free. ISO 13485 and ISO 8 clean room facility.
Packaging:
10 plate/pack; 50 plates/case.
Frequently Bought Together