Order Entry
Austria
ContactUsLinkComponent
SPINeasy® DNA Kit for Microbiome
SPINeasy® DNA Kit for Microbiome
Catalog # 116553050.
Supplier:  MP Biomedicals
New Product
CAS Number:  
SPINeasy® DNA Kit for Microbiome
Catalog # 116553050.
Supplier:  MP Biomedicals
New Product
CAS Number:  

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Sample type:
    Stool
    Swabs
    Biological fluids
    Environmental
  • Target molecule:
    DNA
  • Description:
    SPINeasy® DNA kit for microbiome/gDNA microbial extraction kit
  • Storage temperature:
    Store at room temperature (15...30 °C), dry environment
  • Contents:
    Components: Lysing matrix E (50 ea), RNase A solution (2 ml), buffer MB1 (45 ml), buffer MB2 (17 ml), buffer MB3 (21×2 ml), buffer MB4 (28 ml), buffer MB5 (20×2 ml), buffer MB6 (10 ml), Column MB (50 ea), 1.5 ml collection tubes (50 ea)
  • No. of reactions:
    50 preps

Specifications

About this item

SPINeasy® DNA Kit for Microbiome employs a novel method for isolating microbial genomic DNA from various samples. Due to its highly effective lysis capability and silica-membrane spin-column technology, the SPINeasy® DNA Kit for Microbiome can be used for bacterial, body fluids and environmental samples such as soil and stool.

  • For genomic, molecular biology, PCR, qPCR, microbiome analysis, NGS, Shotgun sequencing

    Personal protective equipment: Safety goggles/face shield, gloves

With the use of specially formulated Buffer MB1 and Lysing Matrix E in combination with FastPrep® Instruments from MP Biomedicals, an efficient lysis of various samples can be achieved within seconds. Provided in the kit, the Column MB and kit buffers are designed to deliver gDNA of high yield and purity, and compatible with downstream applications such as qPCR, restriction digestion, and sequencing.

Efficient and unbiased lysis of microbes, including gram positive/negative bacteria, fungi, protozoans, and viruses found in swab, sputum, body fluid, stool, soil, and water samples.
 

SPINeasy® DNA Kit for Microbiome employs a novel method for isolating microbial genomic DNA from various samples. Due to its highly effective lysis capability and silica membrane spin column technology, the SPINeasy DNA Kit for Microbiome can be used for bacterial, body fluids, and environmental samples such as soil and stool.