Order Entry
Austria
ContactUsLinkComponent
PIG® High Flow Roof Leak Diverter
PIG® High Flow Roof Leak Diverter
Catalog # NEWPTLS676-TR
Supplier:  NEW PIG
New Product
PIG® High Flow Roof Leak Diverter
Catalog # NEWPTLS676-TR
Supplier:  NEW PIG
New Product

Specifications

  • Weight:
    7,6 kg
  • Capacity:
    38 L/minute (flow rate)
  • Description:
    High flow roof leak diverter, square
  • L×W:
    3,4×3,4 m

Specifications

About this item

High flow design catches and diverts up to 38 L per minute to prevent high volume ceiling leaks from becoming a big problem.

  • Easy and quick way to prevent damage to your facility caused by roof or ceiling leaks

    76,2 mm diameter flat hose drains up to 38 L of liquid per minute

    Heavy-duty vinyl-coated tarp resists punctures, UV and mildew and lasts multiple deployments

    Ideal for washdowns or after heavy rains

    Can be installed using hanging straps

Pinched corners create a funnel to channel liquids to a drain or collection container without overflowing or sagging; engineered with leakproof seams and connections. Brass grommets resist tearing and make installation easy; multiple hanging points add stability and preserve funnel shape.

Extra-large drain opening and debris cage prevents clogs from crumbled ceiling tiles, drywall and roofing material. Cut drain hose for a custom fit; insert 90° elbow between hose sections to prevent kinking (hose and elbow fitting included).

Composition:

Diverter - Reinforced vinyl

Hose - Vinyl

Elbow - PVC

Ordering information:

Collection container and hanging straps sold separately.

Delivery:

Includes 0,3 - 3,3W×3,3L m leak diverter tarp, 25 - 76W mm × 7,6L m flat hose, 1 to 90° elbow.