b.safe Exhaust Filter Adapter Justrite
New Product
Adaptor for the use of b.safe Exhaust filters on Justrite® Disposal safety cans in order to stop solvent vapours from the escaping in the air during filling with HPLC waste.
Made of grey PP or black PE
- With GL 14 thread
Adaptor for assembly on the lateral couplings of Justrite® Disposal Safety Cans, with connector to screw in a b.safe Exhaust filter (thread GL 14) for venting the disposal can during filling. Available either in gray PP for a PP-coupling or in black PE for a stainless steel coupling.
Frequently Bought Together